missing translation for 'onlineSavingsMsg'
Learn More

PR-619, MedChemExpress

Product Code. 16415878
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16415878

Brand: Medchem Express HY13814

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PR-619 is a broad-range and reversible DUB inhibitor with EC50s of 3.93, 4.9, 6.86, 7.2, and 8.61 μM for USP4, USP8, USP7, USP2, and USP5, respectively. PR-619 induces ER Stress and ER-Stress related apoptosis.

PR-619 is a broad-range and reversible DUB inhibitor with EC50s of 3.93, 4.9, 6.86, 7.2, and 8.61 μM for USP4, USP8, USP7, USP2, and USP5, respectively. PR-619 induces ER Stress and ER-Stress related apoptosis.

Specifications

Chemical Name or Material PR-619
CAS 2645-32-1
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C7H5N5S2
Quantity 10 mg
Solubility Information DMSO : ≥ 21 mg/mL (94.05 mM)
SMILES NC1=C(SC#N)C=C(SC#N)C(N)=N1
Molecular Weight (g/mol) 223.28
Formula Weight 223.28
Percent Purity 98.04%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.