missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PR-619, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
PR-619 is a broad-range and reversible DUB inhibitor with EC50s of 3.93, 4.9, 6.86, 7.2, and 8.61 μM for USP4, USP8, USP7, USP2, and USP5, respectively. PR-619 induces ER Stress and ER-Stress related apoptosis.
Specifications
Specifications
| Chemical Name or Material | PR-619 |
| CAS | 2645-32-1 |
| Color | Earth Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C7H5N5S2 |
| Quantity | 50 mg |
| Solubility Information | DMSO : ≥ 21 mg/mL (94.05 mM) |
| SMILES | NC1=C(SC#N)C=C(SC#N)C(N)=N1 |
| Molecular Weight (g/mol) | 223.28 |
| Formula Weight | 223.28 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction