missing translation for 'onlineSavingsMsg'
Learn More

PQBP1 Antibody, FITC, Novus Biologicals™

Product Code. 18594993 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18594993 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18594993 Supplier Novus Biologicals Supplier No. NBP147270F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PQBP1 Polyclonal antibody specifically detects PQBP1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PQBP1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias 38 kDa nuclear protein containing a WW domain, MRX55, MRXS3, MRXS8, Npw38, NPW38mental retardation, X-linked 55, nuclear protein containing WW domain 38 kD, polyglutamine binding protein 1, Polyglutamine tract-binding protein 1, polyglutamine-binding protein 1, PQBP-1, RENS1, SHS, Sutherland-Haan X-linked mental retardation syndrome
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 150 and 200 of human Polyglutamine Binding Protein 1 using the numbering given in entry NP_005701.1 (GeneID 10084).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10084
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.