missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPYR1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21654-1-AP-20UL
This item is not returnable.
View return policy
Description
Receptor for neuropeptide Y and peptide YY. The rank order of affinity of this receptor for pancreatic polypeptides is PP, PP (2-36) and [Ile-31, Gln-34] PP > [Pro-34] PYY > PYY and [Leu-31, Pro-34] NPY > NPY > PYY (3-36) and NPY (2-36) > PP (13- 36) > PP (31-36) > NPY free acid.Specifications
| PPYR1 | |
| Polyclonal | |
| Unconjugated | |
| NPY4R | |
| Neuropeptide Y receptor type 4, PP1, PPYR1, Y4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5540 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P50391 | |
| NPY4R | |
| PPYR1 Fusion Protein Ag16374 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction