missing translation for 'onlineSavingsMsg'
Learn More

PPWD1 Antibody (OTI1B8), DyLight 594, Novus Biologicals™

Product Code. 18424217 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424217 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18424217 Supplier Novus Biologicals Supplier No. NBP273590DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PPWD1 Monoclonal antibody specifically detects PPWD1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPWD1
Applications Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI1B8
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias EC 5.2.1.8, KIAA0073Spliceosome-associated cyclophilin, peptidylprolyl isomerase domain and WD repeat containing 1, peptidylprolyl isomerase domain and WD repeat-containing protein 1
Host Species Mouse
Immunogen Full length recombinant protein of human PPWD1 (NP_056157) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23398
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.