missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ PPTC7 Recombinant Protein Antigen

Product Code. 18370520 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18370520 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18370520 Supplier Novus Biologicals™ Supplier No. NBP190653PEP

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human PPTC7. The PPTC7 Recombinant Protein Antigen is derived from E. coli. The PPTC7 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.

This is a blocking peptide for NBP1-90653. This is a human recombinant protein expressed in E. coli with a N-terminal His-ABP tag and purified with IMAC chromatography.

TRUSTED_SUSTAINABILITY

Specifikationer

Gene ID (Entrez) 160760
Species Human
Purification Method Chromatography
Purity >80%
Concentration 0.5mg/mL
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Formulation PBS and 1M Urea, pH 7.4.
For Use With (Application) Blocking/Neutralizing, Control
Gene Symbol PPTC7
Label Type Unlabeled
Molecular Weight (g/mol) 28kDa
Product Type PPTC7
Quantity 0.1 mL
Regulatory Status RUO
Source E.Coli
Specific Reactivity This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP1-90653. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Immunogen LGDIILTATDGLFDNMPDYMILQELKKLKNSNYESIQQTARSIAEQAHELAYDPNYMSPFAQFACDNGLNVRGGKPDDITVLLSIVAEY
Visa mer Visa mindre

For Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.