Learn More
Invitrogen™ PPT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588446
Description
Immunogen sequence: APAPHRASYK PVIVVHGLFD SSYSFRHLLE YINETHPGTV VTVLDLFDGR ESLRPLWEQV QGFREAVVPI MAKAPQGVHL ICYSQGGLVC RALLSVMDDH NVDSFISLSS PQMGQYGDTD YLKWLFPTSM RSNLYRICYS PWGQEFSICN YWHDPHHDDL YLNASSFLAL INGERDHPNA TVWRKNFLRV GHLVLIGGPD DGVITPWQSS FFGFYDANET VLEMEEQLVY LRDSFGLKTL LARGAIVRCP MAGISHTAWH SNRTLYETCI EPWLS; Positive Samples: Mouse liver; Cellular Location: Lysosome.
This gene encodes a member of the palmitoyl-protein thioesterase family. The encoded glycosylated lysosomal protein has palmitoyl-CoA hydrolase activity in vitro, but does not hydrolyze palmitate from cysteine residues in proteins. Three transcript variants encoding different isoforms have been described for this gene, with one of the isoforms being inactive.
Specifications
| PPT2 | |
| Polyclonal | |
| Unconjugated | |
| PPT2 | |
| 0610007M19Rik; AA672937; C6orf8; G14; Lysosomal thioesterase PPT2; palmitoyl-protein hydrolase 2; palmitoyl-protein thioesterase 2; PPT2; PPT-2; S-thioesterase G14; VE-statin2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 54397 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O35448 | |
| PPT2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 28-302 of human PPT2 (NP_0051464). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.