missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPT1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67699-1-IG-20UL
This item is not returnable.
View return policy
Description
The PPT1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PPT1. This antibody recognizes human, pig, rabbit antigen. The PPT1 antibody has been validated for the following applications: WB, IF-P, ELISA analysis.Specifications
| PPT1 | |
| Monoclonal | |
| 1000 μg/mL | |
| CLN1; INCL; PPT | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Pig, Rabbit | |
| 32-34 kDa | |
| Liquid |
| Western Blot, Immunofluorescence, ELISA | |
| 1A1A3 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 5538 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction