missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Protein Precipitation Solution (PPS) for Soil Kit
Shop All MP Biomedicals Products
Click to view available options
For Use With (Equipment):
All FastPrep Adapters
Unit Size:
1mL
Description
- Applications: PPS is component of FastDNA™ SPIN Kit for Soil.
- Usage Statement:
- Research Use Only (RUO).
- Ready for CE IVD labeling of clinical applications.
- All Fast DNA Kit reagents are stable at room temperature(15°C to 30°C).
- Shelf life can be extended by storing at 4°C.
Specifications
Specifications
| For Use With (Application) | Protein Precipitation Solution |
| For Use With (Equipment) | All FastPrep Adapters |
| Packaging Quantity | 25 mL |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction