missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPRC1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25429-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is similar to PPAR-gamma coactivator 1 (PPARGC1/PGC-1), a protein that can activate mitochondrial biogenesis in part through a direct interaction with nuclear respiratory factor 1 (NRF1). This protein has been shown to interact with NRF1. It is thought to be a functional relative of PPARGC1 that activates mitochondrial biogenesis through NRF1 in response to proliferative signals.Specifications
| PPRC1 | |
| Polyclonal | |
| Unconjugated | |
| PPRC1 | |
| KIAA0595, PGC 1 related coactivator, PPRC1, PRC, RP11 302K17.6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 226169, 23082 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5VV67, Q6NZN1 | |
| Pprc1 | |
| PPRC1 Fusion Protein Ag11940 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction