missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PPQ 102

Product Code. p-5545970
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18701789

Brand: Tocris Bioscience™ 4303/50

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent, voltage-independent CFTR chloride channel inhibitor

PPQ 102 is a potent, voltage-independent CFTR chloride channel inhibitor (IC50 = 90 nM). Reduces the size and number of renal cysts in an embryonic kidney culture model of polycystic kidney disease. Also increases VEGF-A production from airway epithelial cells in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Purity >98%
Quantity 50 mg
Product Type PPQ 102
Molecular Weight (g/mol) 438.48
Inhibitors PPQ 102

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.