missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PPPDE1 Polyclonal specifically detects PPPDE1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
| Antigen | PPPDE1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1-2 μg/mL, ELISA 1:100-1:2000 |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Accession No. | NP_057160 |
| Gene Alias | C1orf121, CGI-146, chromosome 1 open reading frame 121, FAM152A, FLJ21998, member A, PNAS-4, PPPDE peptidase domain containing 1, PPPDE peptidase domain-containing protein 1, Protein FAM152A |
| Gene Symbols | DESI2 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?