missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PPPDE1 Polyclonal antibody specifically detects PPPDE1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | PPPDE1 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 750 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | C1orf121, CGI-146, chromosome 1 open reading frame 121, FAM152A, FLJ21998, member A, PNAS-4, PPPDE peptidase domain containing 1, PPPDE peptidase domain-containing protein 1, Protein FAM152A |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human PNAS4. The immunogen is located within the last 50 amino acids of PNAS4. Amino Acid Squence: VSQELQDELEEAEDAAAS |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?