missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP6C Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33542-100ul
This item is not returnable.
View return policy
Description
PPP6C Monoclonal antibody specifically detects PPP6C in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| PPP6C | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| EC 3.1.3.16, FLJ92648, MGC12249, PP6, PP6C, PPP6, protein phosphatase 6, catalytic subunit, serine/threonine protein phosphatase catalytic subunit, serine/threonine-protein phosphatase 6 catalytic subunit | |
| A synthetic peptide corresponding to a sequence within amino acids 206-305 of human PPP6C (O00743).,, Sequence:, AISPRGAGWLFGAKVTNEFVHINNLKLICRAHQLVHEGYKFMFDEKLVTVWSAPNYCYRCGNIASIMVFKDVNTREPKLFRAVPDSERVIPPRTTTPYFL | |
| 100 μL | |
| Cell Cycle and Replication, Signal Transduction | |
| 5537 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction