missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPP4R4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532166
This item is not returnable.
View return policy
Description
Recommended positive controls: U87-MG. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a HEAT-like repeat-containing protein. The HEAT repeat is a tandemly repeated, 37-47 amino acid long module occurring in a number of cytoplasmic proteins. Arrays of HEAT repeats form a rod-like helical structure and appear to function as protein-protein interaction surfaces. The repeat-containing region of this protein has some similarity to the constant regulatory domain of the protein phosphatase 2A PR65/A subunit. The function of this particular gene product has not been determined. Alternative splicing has been observed for this gene and two transcript variants encoding distinct isoforms have been identified.
Specifications
| PPP4R4 | |
| Polyclonal | |
| Unconjugated | |
| PPP4R4 | |
| 8430415E04Rik; CFAP14; cilia and flagella associated protein 14; HEAT-like repeat-containing protein; KIAA1622; mKIAA1622; PP4R4; PPP4R4; protein phosphatase 4 regulatory subunit 4; protein phosphatase 4, regulatory subunit 4; RGD1560932; Serine/threonine-protein phosphatase 4 regulatory subunit 4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57718 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q6NUP7 | |
| PPP4R4 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human PP4R4. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction