Learn More
Novus Biologicals™ PPP4R1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-30785-100ug
 View more versions of this product
Product Code. 30067187
  • £386.00 / 100µg
Description
PPP4R1 Polyclonal antibody specifically detects PPP4R1 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PPP4R1
Polyclonal
Phosphate buffered saline
Rabbit
Ammonium sulfate precipitation
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot
Unconjugated
MEG1, PP4R1PP4(Rmeg), protein phosphatase 4, regulatory subunit 1, serine/threonine phosphatase 4, serine/threonine-protein phosphatase 4 regulatory subunit 1
Antibody raised against synthetic peptide corresponding to amino acids 38 to 52 of the human protein PPP4R1.
100 μg
Protein Phosphatase
9989
Store at -20°C. Avoid freeze/thaw cycles.