missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP4R1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£149.00 - £366.00
Specifications
| Antigen | PPP4R1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, Immunoprecipitation 1:50 - 1:100 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18668431
|
Novus Biologicals
NBP2-94582-0.02ml |
0.02 mL |
£149.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18627541
|
Novus Biologicals
NBP2-94582-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PPP4R1 Polyclonal antibody specifically detects PPP4R1 in Human, Mouse samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| PPP4R1 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Protein Phosphatase | |
| PBS (pH 7.3), 50% glycerol | |
| 9989 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, Immunoprecipitation 1:50 - 1:100 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| MEG1, PP4R1PP4(Rmeg), protein phosphatase 4, regulatory subunit 1, serine/threonine phosphatase 4, serine/threonine-protein phosphatase 4 regulatory subunit 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 324-647 of human PPP4R1 (NP_001035847.1). NPSSSGQYFKEESKSSEEMSVENKNRTRDQEAPEDVQVRPEDTPSDLSVSNSSVILENTMEDHAAEASGKPLGEISVPLDSSLLCTLSSESHQEAASNENDKKPGNYKSMLRPEVGTTSQDSALLDQELYNSFHFWRTPLPEIDLDIELEQNSGGKPSPEGPEEESEGPVPSSPNITMATRKELEEMIENLEPHIDDPDVKAQVEVLSAALRASSLDAHEETISIEKRSDLQDELDINELPNCKINQEDSVPLISDAVENMDSTLHYIHSDSDLSNNSSFSPDEERRTKVQDVVPQALLDQYLSMTDPSRAQTVDTEIAKHCAY | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title