missing translation for 'onlineSavingsMsg'
Learn More

PPP3R2 Antibody - BSA Free, Novus Biologicals™

Product Code. p-200060760 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.10mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18603231 0.02 mL 0.02mL
18607210 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18603231 Supplier Novus Biologicals Supplier No. NBP2932110.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PPP3R2 Polyclonal antibody specifically detects PPP3R2 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP3R2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias calcineurin B, type II (19kDa), Calcineurin BII, Calcineurin B-like protein, calcineurin subunit B type 2, CBLP, CNBII, PPP3RL, Protein phosphatase 2B regulatory subunit 2, protein phosphatase 3 (formerly 2B), regulatory subunit B (19kD), beta isoform(calcineurin B, type II), protein phosphatase 3 (formerly 2B), regulatory subunit B, 19kDa, beta isoform(calcineurin B, type II), protein phosphatase 3 (formerly 2B), regulatory subunit B, beta isoform, Protein phosphatase 3 regulatory subunit B beta isoform, protein phosphatase 3, regulatory subunit B, beta
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-173 of human PPP3R2 (NP_671709.1). MSTMGNEASYPAEMCSHFDNDEIKRLGRRFKKLDLDKSGSLSVEEFMSLPELRHNPLVRRVIDVFDTDGDGEVDFKEFILGTSQFSVKGDEEQKLRFAFSIYDMDKDGYISNGELFQVLKMMVGNNLTDWQLQQLVDKTIIILDKDGDGKISFEEFSAVVRDLEIHKKLVLIV
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5535
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.