missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP3R2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93211-0.1ml
This item is not returnable.
View return policy
Description
PPP3R2 Polyclonal antibody specifically detects PPP3R2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PPP3R2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| calcineurin B, type II (19kDa), Calcineurin BII, Calcineurin B-like protein, calcineurin subunit B type 2, CBLP, CNBII, PPP3RL, Protein phosphatase 2B regulatory subunit 2, protein phosphatase 3 (formerly 2B), regulatory subunit B (19kD), beta isoform(calcineurin B, type II), protein phosphatase 3 (formerly 2B), regulatory subunit B, 19kDa, beta isoform(calcineurin B, type II), protein phosphatase 3 (formerly 2B), regulatory subunit B, beta isoform, Protein phosphatase 3 regulatory subunit B beta isoform, protein phosphatase 3, regulatory subunit B, beta | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-173 of human PPP3R2 (NP_671709.1). MSTMGNEASYPAEMCSHFDNDEIKRLGRRFKKLDLDKSGSLSVEEFMSLPELRHNPLVRRVIDVFDTDGDGEVDFKEFILGTSQFSVKGDEEQKLRFAFSIYDMDKDGYISNGELFQVLKMMVGNNLTDWQLQQLVDKTIIILDKDGDGKISFEEFSAVVRDLEIHKKLVLIV | |
| 0.1 mL | |
| Wnt Signaling Pathway | |
| 5535 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction