missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PPP3R2 Polyclonal antibody specifically detects PPP3R2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | PPP3R2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | calcineurin B, type II (19kDa), Calcineurin BII, Calcineurin B-like protein, calcineurin subunit B type 2, CBLP, CNBII, PPP3RL, Protein phosphatase 2B regulatory subunit 2, protein phosphatase 3 (formerly 2B), regulatory subunit B (19kD), beta isoform(calcineurin B, type II), protein phosphatase 3 (formerly 2B), regulatory subunit B, 19kDa, beta isoform(calcineurin B, type II), protein phosphatase 3 (formerly 2B), regulatory subunit B, beta isoform, Protein phosphatase 3 regulatory subunit B beta isoform, protein phosphatase 3, regulatory subunit B, beta |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-173 of human PPP3R2 (NP_671709.1). MSTMGNEASYPAEMCSHFDNDEIKRLGRRFKKLDLDKSGSLSVEEFMSLPELRHNPLVRRVIDVFDTDGDGEVDFKEFILGTSQFSVKGDEEQKLRFAFSIYDMDKDGYISNGELFQVLKMMVGNNLTDWQLQQLVDKTIIILDKDGDGKISFEEFSAVVRDLEIHKKLVLIV |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?