Learn More
PPP3R1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13210-1-AP-150UL
 View more versions of this product
Product Code. 16871314
  • £349.00 / 150µL
Description
Protein phosphatase 3 (PPP3, formerly PP2B, Calcineurin), a serine/ threonine protein phosphatase, is a heterodimer composed of one catalytic subunit (PPP3C, Calcineurin A) and one regulatory subunit (PPP3R, Calcineurin B). PPP3R has two isoforms, PPP3R1 and PPP3R2. PPP3Rl is ubiquitously expressed in different tissues and is coded by the PPP3R1 gene which was mapped recently to human chromosome 2. PPP3Rl has four functional calcium-binding sites.
Specifications
Specifications
PPP3R1
Polyclonal
Unconjugated
PPP3R1
Calcineurin subunit B type 1, CALNB1, CNA2, CNB, CNB1, PPP3R1
Rabbit
Antigen Affinity Chromatography
RUO
5534
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P63098
PPP3R1
PPP3R1 Fusion Protein Ag3992
150 μL