missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP3CB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13340-1-AP-20UL
This item is not returnable.
View return policy
Description
PPP3CB is a calcium-dependent, calmodulin-stimulated protein phosphatase. This subunit may have a role in the calmodulin activation of calcineurin.Specifications
| PPP3CB | |
| Polyclonal | |
| Unconjugated | |
| PPP3CB | |
| CALNA2, CALNB, CAM PRP catalytic subunit, CNA2, PPP3CB | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19056, 24675, 5532 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P16298, P20651, P48453 | |
| Ppp3cb | |
| PPP3CB Fusion Protein Ag4163 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction