missing translation for 'onlineSavingsMsg'
Learn More

PPP3CB Antibody (OTI2E4) [Janelia Fluor™ 669], Novus Biologicals™

Product Code. 30688731 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30688731 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30688731 Supplier Novus Biologicals Supplier No. NBP345023JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PPP3CB Monoclonal antibody specifically detects PPP3CB in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP3CB
Applications Western Blot
Classification Monoclonal
Clone OTI2E4
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias Calmodulin-dependent calcineurin A subunit beta isoform, CALNA2protein phosphatase 3 (formerly 2B), catalytic subunit, beta isoform(calcineurin A beta), CAM-PRP catalytic subunit, CNA2catalytic subunit, beta isoform, EC 3.1.3.16, protein phosphatase 2B, catalytic subunit, beta isoform, protein phosphatase 3, catalytic subunit, beta isozyme, protein phosphatase from PCR fragment H32, serine/threonine-protein phosphatase 2B catalytic subunit beta isoform
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 23-96 of human PPP3CB (NP_001135826) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5532
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.