missing translation for 'onlineSavingsMsg'
Learn More

PPP3CB Antibody (OTI2E4) [Alexa Fluor™ 750], Novus Biologicals™

Product Code. 30687780 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30687780 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30687780 Supplier Novus Biologicals Supplier No. NBP345023AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PPP3CB Monoclonal antibody specifically detects PPP3CB in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP3CB
Applications Western Blot
Classification Monoclonal
Clone OTI2E4
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias Calmodulin-dependent calcineurin A subunit beta isoform, CALNA2protein phosphatase 3 (formerly 2B), catalytic subunit, beta isoform(calcineurin A beta), CAM-PRP catalytic subunit, CNA2catalytic subunit, beta isoform, EC 3.1.3.16, protein phosphatase 2B, catalytic subunit, beta isoform, protein phosphatase 3, catalytic subunit, beta isozyme, protein phosphatase from PCR fragment H32, serine/threonine-protein phosphatase 2B catalytic subunit beta isoform
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 23-96 of human PPP3CB (NP_001135826) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5532
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.