missing translation for 'onlineSavingsMsg'
Learn More

PPP2R5E Antibody, DyLight 680, Novus Biologicals™

Product Code. 18370593 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18370593 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18370593 Supplier Novus Biologicals Supplier No. NBP306478FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PPP2R5E Polyclonal antibody specifically detects PPP2R5E in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP2R5E
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias epsilon isoform of regulatory subunit B56, protein phosphatase 2A, PP2A B subunit isoform B56-epsilon, PP2A B subunit isoform B'-epsilon, PP2A B subunit isoform PR61-epsilon, PP2A B subunit isoform R5-epsilon, PP2A, B subunit, B' epsilon, PP2A, B subunit, B56 epsilon, PP2A, B subunit, PR61 epsilon, PP2A, B subunit, R5 epsilon, protein phosphatase 2, regulatory subunit B (B56), epsilon isoform, protein phosphatase 2, regulatory subunit B', epsilon isoform, serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, epsilon, serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit epsilonisoform
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human PPP2R5E fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5529
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.