missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PPP2R5B Polyclonal Antibody
GREENER_CHOICE

Product Code. 13259998
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13259998 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13259998 Supplier Invitrogen™ Supplier No. PA521763

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: A431. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes the phosphatase 2A catalytic subunit. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. This gene encodes a beta isoform of the catalytic subunit. Two transcript variants encoding the same protein have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP2R5B
Applications Western Blot
Classification Polyclonal
Concentration 0.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 10% glycerol and 0.01% thimerosal; pH 7
Gene PPP2R5B
Gene Accession No. Q15173
Gene Alias B56B; B'beta; BC026670; FLJ35411; LOC100909468; PP2A; PP2A B subunit isoform B56-beta; PP2A B subunit isoform B'-beta; PP2A B subunit isoform PR61-beta; PP2A B subunit isoform R5-beta; Ppp2r5b; PR61B; protein phosphatase 2 regulatory subunit B', beta; protein phosphatase 2 regulatory subunit B'beta; protein phosphatase 2, regulatory subunit B (B56), beta isoform; protein phosphatase 2, regulatory subunit B', beta; protein phosphatase 2, regulatory subunit B', beta isoform; protein phosphotase 2, regulatory subunit B (B56), beta isoform; rCG_47223; serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, beta isoform; serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit beta isoform
Gene Symbols PPP2R5B
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human PPP2R5B. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5526
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.