missing translation for 'onlineSavingsMsg'
Learn More

PPP2R5B Antibody, Novus Biologicals™

Product Code. 18357778 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18357778 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18357778 Supplier Novus Biologicals Supplier No. H00005526B01P50ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

PPP2R5B Polyclonal antibody specifically detects PPP2R5B in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP2R5B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_006235.1
Gene Alias B56B, FLJ35411, PP2A B subunit isoform B56-beta, PP2A B subunit isoform B'-beta, PP2A B subunit isoform PR61-beta, PP2A B subunit isoform R5-beta, PR61B, protein phosphatase 2, regulatory subunit B (B56), beta isoform, protein phosphatase 2, regulatory subunit B', beta, protein phosphatase 2, regulatory subunit B', beta isoform, serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, betaisoform, serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit beta isoform
Host Species Mouse
Immunogen PPP2R5B (NP_006235.1, 1 a.a. - 497 a.a.) full-length human protein. METKLPPASTPTSPSSPGLSPVPPPDKVDGFSRRSLRRARPRRSHSSSQFRYQSNQQELTPLPLLKDVPASELHELLSRKLAQCGVMFDFLDCVADLKGKEVKRAALNELVECVGSTRGVLIEPVYPDIIRMISVNIFRTLPPSENPEFDPEEDEPNLEPSWPHLQLVYEFFLRFLESPDFQPSVAKRYVDQKFVLMLLELFDSEDPREREYLKTILHRVYGKFLGLRAYIRKQCNHIFLRFIYEFEHFNGVAELLEILGSIINGFALPLKTEHKQFLVRVLIPLHSVKSLSVFHAQLAYCVVQFLEKDATLTEHVIRGLLKYWPKTCTQKEVMFLGEMEEILDVIEPSQFVKIQEPLFKQVARCVSSPHFQVAERALYFWNNEYILSLIEDNCHTVLPAVFGTLYQVSKEHWNQTIVSLIYNVLKTFMEMNGKLFDELTASYKLEKQQEQQKAQERQELWQGLEELRLRRLQGTQGAKEAPLQRLTPQVAASGGQS
Purification Method Immunogen affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5526
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.