missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP2R5A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12675-2-AP-20UL
This item is not returnable.
View return policy
Description
The product of this gene belongs to the phosphatase 2A regulatory subunit B family. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The B regulatory subunit might modulate substrate selectivity and catalytic activity. This gene encodes an alpha isoform of the regulatory subunit B56 subfamily.Specifications
| PPP2R5A | |
| Polyclonal | |
| Unconjugated | |
| PPP2R5A | |
| B56A, PP2A B subunit isoform B alpha, PPP2R5A, PR61A, PR61alpha | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 226849, 312754, 5525 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q15172 | |
| Ppp2r5a | |
| PPP2R5A Fusion Protein Ag3360 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction