missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PPP2R5A Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30774-100ug
This item is not returnable.
View return policy
Description
PPP2R5A Polyclonal antibody specifically detects PPP2R5A in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| PPP2R5A | |
| Polyclonal | |
| Western Blot : 5-10 μg/mL, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence : 2-10 μg/mL | |
| B56A, MGC131915, PP2A B subunit isoform B56-alpha, PP2A B subunit isoform B'-alpha, PP2A B subunit isoform PR61-alpha, PP2A B subunit isoform R5-alpha, PP2A, B subunit, B' alpha isoform, PP2A, B subunit, B56 alpha isoform, PP2A, B subunit, PR61 alpha isoform, PP2A, B subunit, R5 alpha isoform, PR61A, PR61alpha, protein phosphatase 2, regulatory subunit B (B56), alpha isoform, protein phosphatase 2, regulatory subunit B', alpha, protein phosphatase 2, regulatory subunit B', alpha isoform, serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, alphaisoform, serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit alpha isoform | |
| A peptide conjugated to KLH | |
| 100 μg | |
| Wnt Signaling Pathway | |
| 5525 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Phosphate buffered saline | |
| Sheep | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction