missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ PPP2R5A Antibody, Novus Biologicals™

Sheep Polyclonal Antibody

Brand:  Novus Biologicals™ NBP3-30774-25ug

 View more versions of this product

Product Code. 30067091

  • £162.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PPP2R5A Polyclonal antibody specifically detects PPP2R5A in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PPP2R5A
Polyclonal
Western Blot : 5-10 μg/mL, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence : 2-10 μg/mL
B56A, MGC131915, PP2A B subunit isoform B56-alpha, PP2A B subunit isoform B'-alpha, PP2A B subunit isoform PR61-alpha, PP2A B subunit isoform R5-alpha, PP2A, B subunit, B' alpha isoform, PP2A, B subunit, B56 alpha isoform, PP2A, B subunit, PR61 alpha isoform, PP2A, B subunit, R5 alpha isoform, PR61A, PR61alpha, protein phosphatase 2, regulatory subunit B (B56), alpha isoform, protein phosphatase 2, regulatory subunit B', alpha, protein phosphatase 2, regulatory subunit B', alpha isoform, serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, alphaisoform, serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit alpha isoform
A peptide conjugated to KLH
25 μg
Wnt Signaling Pathway
5525
Store at -20°C. Avoid freeze/thaw cycles.
IgG
Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
Unconjugated
Phosphate buffered saline
Sheep
Ammonium sulfate precipitation
RUO
Primary
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.