Learn More
Invitrogen™ PPP2R4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595986
Description
Immunogen sequence: MAEGERQPPP DSSEEAPPAT QNFIIPKKEI HTVPDMGKWK RSQAYADYIG FILTLNEGVK GKKLTFEYRV SEAIEKLVAL LNTLDRWIDE TPPVDQPSRF GNKAYRTWYA KLDEEAENLV ATVVPTHLAA AVPEVAVYLK ESVGNSTRID YGTGHEAAFA AFLCCLCKIG VLRVDDQIAI VFKVFNRYLE VMRKLQKTYR MEPAGSQGVW GLDDFQFLPF IWGSSQLIDH PYLEPRHFVD EKAVNENHKD YMFLECILFI TEMKTGPFAE HSNQLWNISA VPSWSKVNQG LIRMYKAECL EKFPVIQHFK FGSLLPIHPV TSG.
Protein phosphatase 2A is one of the four major Ser/Thr phosphatases and is implicated in the negative control of cell growth and division. Protein phosphatase 2A holoenzymes are heterotrimeric proteins composed of a structural subunit A, a catalytic subunit C, and a regulatory subunit B. The regulatory subunit is encoded by a diverse set of genes that have been grouped into the B/PR55, B'/PR61, and B'/PR72 families. These different regulatory subunits confer distinct enzymatic specificities and intracellular localizations to the holozenzyme. The product of this gene belongs to the B' family. This gene encodes a specific phosphotyrosyl phosphatase activator of the dimeric form of protein phosphatase 2A. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| PPP2R4 | |
| Polyclonal | |
| Unconjugated | |
| PTPA | |
| 2610042B21Rik; C77440; MGC2184; N28142; OTTHUMP00000022333; phosphotyrosyl phosphatase activator; PP2A; PP2A phosphatase activator; PP2A subunit B' isoform PR53; PP2A, subunit B', PR53 isoform; Ppp2r4; PR53; protein phosphatase 2 phosphatase activator; protein phosphatase 2 regulatory subunit 4; protein phosphatase 2A activator, regulatory subunit 4; protein phosphatase 2A activator, regulatory subunit B; protein phosphatase 2A regulatory subunit 4; protein phosphatase 2A, regulatory subunit B (PR 53); protein phosphatase 2A, regulatory subunit B' (PR 53); PTPA; Serine/threonine-protein phosphatase 2A activator; serine/threonine-protein phosphatase 2A regulatory subunit 4; Serine/threonine-protein phosphatase 2A regulatory subunit B' | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 110854, 5524 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P58389, Q15257 | |
| PTPA | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-323 of human Ser/thr-PP2A activator (PTPA) (NP_8210671). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.