missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP2R3A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-54581
This item is not returnable.
View return policy
Description
PPP2R3A Polyclonal specifically detects PPP2R3A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| PPP2R3A | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| PP2A subunit B isoform R3 isoform, PP2A subunit B isoforms B72/B130, PP2A subunit B isoforms B'-PR72/PR130, protein phosphatase 2 (formerly 2A), regulatory subunit B' (PR 72), alphaisoform and (PR 130), beta isoform, protein phosphatase 2 (formerly 2A), regulatory subunit B', alpha, protein phosphatase 2, regulatory subunit B', alpha, serine/threonine-protein phosphatase 2A regulatory subunit B' subunit alpha, subunit B, R3 isoform | |
| Rabbit | |
| 130 kDa | |
| 100 μL | |
| Primary | |
| This product is specific to Subunit or Isoform: B. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Q06190 | |
| PPP2R3A | |
| Synthetic peptides corresponding to PPP2R3A(protein phosphatase 2 (formerly 2A), regulatory subunit B'', alpha) The peptide sequence was selected from the N terminal of PPP2R3A. Peptide sequence SSSVEEKPLSHRNSLDTNLTSMFLQNFSEEDLVTQILEKHKIDNFSSGTD The peptide sequence for this immunogen was taken from within the described region. | |
| Affinity purified | |
| RUO | |
| 5523 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction