missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP2R3A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | PPP2R3A |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18263403
|
Novus Biologicals
NBP2-55833 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18610329
|
Novus Biologicals
NBP2-55833-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PPP2R3A Polyclonal specifically detects PPP2R3A in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| PPP2R3A | |
| Polyclonal | |
| Rabbit | |
| Human | |
| PP2A subunit B isoform R3 isoform, PP2A subunit B isoforms B72/B130, PP2A subunit B isoforms B'-PR72/PR130, protein phosphatase 2 (formerly 2A), regulatory subunit B' (PR 72), alphaisoform and (PR 130), beta isoform, protein phosphatase 2 (formerly 2A), regulatory subunit B', alpha, protein phosphatase 2, regulatory subunit B', alpha, serine/threonine-protein phosphatase 2A regulatory subunit B' subunit alpha, subunit B, R3 isoform | |
| PPP2R3A | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 5523 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:FSEEDLVTQILEKHKIDNFSSGTDIKMCLDILLKCSEDLKKCTDIIKQCIKKKSGSSISEGSGNDTISSSETVYMNVMTRLA | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title