missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPP2R2D Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560112
This item is not returnable.
View return policy
Description
Immunogen sequence: FEEPEDPSSR SFFSEIISSI SDVKFSHSGR YM Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
PPP2R2D is the B regulatory subunit might modulate substrate selectivity and catalytic activity, and also might direct the localization of the catalytic enzyme to a particular subcellular compartment.
Specifications
| PPP2R2D | |
| Polyclonal | |
| Unconjugated | |
| PPP2R2D | |
| 1300017E19Rik; D7Ertd753e; Kiaa1541; MDS026; PP2A subunit B isoform B55-delta; PP2A subunit B isoform delta; PP2A subunit B isoform PR55-delta; PP2A subunit B isoform R2-delta; PP2A, subunit B, B55-delta isoform; PP2A, subunit B, B-delta isoform; PP2A, subunit B, PR55-delta isoform; PP2A, subunit B, R2-delta isoform; PPP2R2D; protein phosphatase 2 regulatory subunit B, delta; protein phosphatase 2 regulatory subunit Bdelta; protein phosphatase 2, regulatory subunit B, delta; protein phosphatase 2, regulatory subunit B, delta isoform; protein phosphatase 2A B regulatory subunit delta isoform; protein phosphatase 2A, regulatory subunit B, delta isoform; serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B delta isoform | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55844 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q66LE6 | |
| PPP2R2D | |
| Recombinant protein corresponding to Human PPP2R2D. Recombinant protein control fragment (Product #RP-98930). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction