missing translation for 'onlineSavingsMsg'
Learn More

PPP2R2D Antibody (GT577), Novus Biologicals™

Product Code. 18353775 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18353775 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18353775 Supplier Novus Biologicals Supplier No. NBP313614

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PPP2R2D Monoclonal antibody specifically detects PPP2R2D in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP2R2D
Applications Western Blot
Classification Monoclonal
Clone GT577
Conjugate Unconjugated
Formulation PBS
Gene Alias KIAA1541, MDS026, PP2A subunit B isoform B55-delta, PP2A subunit B isoform delta, PP2A subunit B isoform PR55-delta, PP2A subunit B isoform R2-delta, protein phosphatase 2, regulatory subunit B, delta, protein phosphatase 2, regulatory subunit B, delta isoform, serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B deltaisoform
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of human PPP2R2D. The exact sequence is proprietary.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 55844
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.