missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP2R2C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12747-1-AP-150UL
This item is not returnable.
View return policy
Description
The B regulatory subunit might modulate substrate selectivity and catalytic activity, and also might direct the localization of the catalytic enzyme to a particular subcellular compartment.Specifications
| PPP2R2C | |
| Polyclonal | |
| Unconjugated | |
| PPP2R2C | |
| B55 GAMMA, IMYPNO, IMYPNO1, PP2A subunit B isoform gamma, PPP2R2C, PR52, PR55G | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 117256, 269643, 5522 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P97888, Q8BG02, Q9Y2T4 | |
| PPP2R2C | |
| PPP2R2C Fusion Protein Ag3470 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction