missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP2R2C Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00005522-B01P
This item is not returnable.
View return policy
Description
PPP2R2C Polyclonal antibody specifically detects PPP2R2C in Human samples. It is validated for Western Blot
Specifications
| PPP2R2C | |
| Polyclonal | |
| Western Blot | |
| AAH32954.1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| B55-GAMMA, IMYPNO1, phosphoprotein phosphatase 2A BR gamma regulatory chain, PP2A subunit B isoform B55-gamma, PP2A subunit B isoform gamma, PP2A subunit B isoform PR55-gamma, PP2A subunit B isoform R2-gamma, PP2A, subunit B, B55-gamma isoform, PP2A, subunit B, B-gamma isoform, PP2A, subunit B, PR55-gamma isoform, PP2A, subunit B, R2-gamma isoform, PR55G, protein phosphatase 2, protein phosphatase 2 (formerly 2A), regulatory subunit B (PR 52), gammaisoform, protein phosphatase 2 (formerly 2A), regulatory subunit B, gamma isoform, protein phosphatase 2, regulatory subunit B, gamma, protein phosphatase 2A1 B gamma subunit, serine/threonine protein phosphatase 2A, 55 KDA regulatory subunit B, gammaisoform, serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B gammaisoform | |
| PPP2R2C (AAH32954.1, 1 a.a. - 430 a.a.) full-length human protein. MGKRDTADIISTVEFNHTGELLATGDKGGRVVIFQREPESKNAPHSQGEYDVYSTFQSHEPEFDYLKSLEIEEKINKIKWLPQQNAAHSLLSTNDKTIKLWKITERDKRPEGYNLKDEEGKLKDLSTVTSLQVPVLKPMDLMVEVSPRRIFANGHTYHINSISVNSDCETYMSADDLRINLWHLAITDRSFNIVDIKPANMEDLTEVITASEFHPHHCNLFVYSSSKGSLRLCDMRAAALCDKHSKLFEEPEDPSNRSFFSEIISSVSDVKFSHSGRYMLTRDYLTVKVWDLNMEARPIETYQVHDYLRSKLCSLYENDCIFDKFECAWNGSDSVIMTGAYNNFFRMFDRNTKRDVTLEASRESSKPRAVLKPRRVCVGGKRRRDDISVDSLDFTKKILHTAWHPAENIIAIAATNNLYIFQDKVNSDMH | |
| 0.05 mg | |
| Protein Phosphatase | |
| 5522 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction