missing translation for 'onlineSavingsMsg'
Learn More

PPP2R2C Antibody (6D1), Novus Biologicals™

Product Code. 18393899 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18393899 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18393899 Supplier Novus Biologicals Supplier No. H00005522M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PPP2R2C Monoclonal antibody specifically detects PPP2R2C in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP2R2C
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6D1
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation In 1x PBS, pH 7.4
Gene Alias B55-GAMMA, IMYPNO1, phosphoprotein phosphatase 2A BR gamma regulatory chain, PP2A subunit B isoform B55-gamma, PP2A subunit B isoform gamma, PP2A subunit B isoform PR55-gamma, PP2A subunit B isoform R2-gamma, PP2A, subunit B, B55-gamma isoform, PP2A, subunit B, B-gamma isoform, PP2A, subunit B, PR55-gamma isoform, PP2A, subunit B, R2-gamma isoform, PR55G, protein phosphatase 2, protein phosphatase 2 (formerly 2A), regulatory subunit B (PR 52), gammaisoform, protein phosphatase 2 (formerly 2A), regulatory subunit B, gamma isoform, protein phosphatase 2, regulatory subunit B, gamma, protein phosphatase 2A1 B gamma subunit, serine/threonine protein phosphatase 2A, 55 KDA regulatory subunit B, gammaisoform, serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B gammaisoform
Host Species Mouse
Immunogen PPP2R2C (NP_065149, 1 a.a. ~ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MGEDTDTRKINHSFLRDHSYVTEADIISTVEFNHTGELLATGDKGGRVVIFQREPESKNAPHSQGEYDVYSTFQSHEPEFDYLKSLEIEEKINKIKWLPQQNAAHSLLST
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 5522
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.