Learn More
PPP2R2B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13123-1-AP-20UL
 View more versions of this product
Product Code. 16881184
  • £135.00 / 20µL
Description
The product of this gene belongs to the phosphatase 2 regulatory subunit B family. Protein phosphatase 2 is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The B regulatory subunit might modulate substrate selectivity and catalytic activity. This gene encodes a beta isoform of the regulatory subunit B55 subfamily. Defects in this gene cause autosomal domit spinocerebellar ataxia 12 (SCA12), a disease caused by degeneration of the cerebellum, sometimes involving the brainstem and spinal cord, and in resulting in poor coordination of speech and body movements. Multiple alternatively spliced variants, which encode different isoforms, have been identified for this gene. The 5′ UTR of some of these variants includes a CAG trinucleotide repeat sequence (7-28 copies) that can be expanded to 66-78 copies in cases of SCA12.
Specifications
Specifications
PPP2R2B
Polyclonal
Unconjugated
PPP2R2B
B55 BETA, PP2A B55BETA, PP2A PR55B, PP2A subunit B isoform beta, PP2A subunit B isoform R2 beta, PP2AB BETA, PP2APR55 BETA, PPP2R2B, PR2AB BETA, PR2AB55 BETA, PR2APR55 BETA, PR52B, PR55 BETA, SCA12
Rabbit
Antigen Affinity Chromatography
RUO
5521, 60660, 72930
-20°C
Liquid
Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry
0.14 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P36877, Q00005, Q6ZWR4
PPP2R2B
PPP2R2B Fusion Protein Ag3860
20 μL
Primary
Rat, Mouse, Human