missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP2R2A Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33476-100ul
This item is not returnable.
View return policy
Description
PPP2R2A Monoclonal antibody specifically detects PPP2R2A in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| PPP2R2A | |
| Monoclonal | |
| Western Blot 1:1000 - 1:5000, ELISA Recommended starting concentration is 1 μg/mL | |
| B55A, B55ALPHA, DKFZp686N05117, FLJ26613, FLJ41727, MGC52248, PP2A subunit B isoform alpha, PP2A subunit B isoform B55-alpha, PP2A subunit B isoform PR55-alpha, PP2A subunit B isoform R2-alpha, PR52A, PR55A, protein phosphatase 2 (formerly 2A), regulatory subunit B (PR 52), alphaisoform, protein phosphatase 2 (formerly 2A), regulatory subunit B (PR52), alpha isoform, protein phosphatase 2 (formerly 2A), regulatory subunit B, alpha isoform, protein phosphatase 2, regulatory subunit B, alpha, serine/threonine protein phosphatase 2A, 55 KDA regulatory subunit B, alphaisoform, serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B alphaisoform | |
| A synthetic peptide corresponding to a sequence within amino acids 94-194 of human PPP2R2A (NP_002708.1).,, Sequence:, EKINKIRWLPQKNAAQFLLSTNDKTIKLWKISERDKRPEGYNLKEEDGRYRDPTTVTTLRVPVFRPMDLMVEASPRRIFANAHTYHINSISINSDYETYLS | |
| 100 μL | |
| Cell Biology, Cell Cycle and Replication, Neuroscience, Signal Transduction | |
| 5520 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction