missing translation for 'onlineSavingsMsg'
Learn More

PPP2R1B Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18327001 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18327001 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18327001 Supplier Novus Biologicals Supplier No. NBP2855150.1ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PPP2R1B Polyclonal specifically detects PPP2R1B in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP2R1B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 2% Sucrose
Gene Alias MGC26454, PP2A subunit A isoform PR65-beta, PP2A subunit A isoform R1-beta, PP2A, subunit A, PR65-beta isoform, PP2A, subunit A, R1-beta isoform, PP2A-Abeta, PR65B, protein phosphatase 2 (formerly 2A), regulatory subunit A (PR 65), beta isoform, protein phosphatase 2 (formerly 2A), regulatory subunit A, beta isoform, protein phosphatase 2, regulatory subunit A, beta, protein phosphatase 2, structural/regulatory subunit A, beta, serine/threonine-protein phosphatase 2A 65 kDa regulatory subunit A betaisoform
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the middle region of human PPP2R1B
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Protein Phosphatase, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5519
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.