missing translation for 'onlineSavingsMsg'
Learn More

PPP2R1A Antibody, DyLight 350, Novus Biologicals™

Product Code. 18420669 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420669 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420669 Supplier Novus Biologicals Supplier No. NBP299603UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PPP2R1A Polyclonal antibody specifically detects PPP2R1A in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP2R1A
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias Medium tumor antigen-associated 61 kDa protein, MGC786, PP2A subunit A isoform PR65-alpha, PP2A subunit A isoform R1-alpha, PP2A-Aalpha, PP2AAALPHA, PR65A, protein phosphatase 2 (formerly 2A), regulatory subunit A (PR 65), alphaisoform, protein phosphatase 2 (formerly 2A), regulatory subunit A, alpha isoform, protein phosphatase 2, regulatory subunit A, alpha, serine/threonine protein phosphatase 2A, 65 kDa regulatory subunit A, alphaisoform, serine/threonine-protein phosphatase 2A 65 kDa regulatory subunit A alphaisoform
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human PPP2R1A.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 5518
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.