missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP2CB Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94074-0.1ml
This item is not returnable.
View return policy
Description
PPP2CB Polyclonal antibody specifically detects PPP2CB in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| PPP2CB | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:100-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin | |
| EC 3.1.3.16, PP2Abeta, PP2A-beta, PP2CB, protein phosphatase 2 (formerly 2A), catalytic subunit, beta isoform, protein phosphatase 2, catalytic subunit, beta isoform, protein phosphatase 2, catalytic subunit, beta isozyme, protein phosphatase 2A catalytic subunit, beta isoform, protein phosphatase type 2A catalytic subunit, serine/threonine protein phosphatase 2A, catalytic subunit, beta isoform, serine/threonine-protein phosphatase 2A catalytic subunit beta isoform | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-309 of human PPP2CB (NP_001009552.1). MDDKAFTKELDQWVEQLNECKQLNENQVRTLCEKAKEILTKESNVQEVRCPVTVCGDVHGQFHDLMELFRIGGKSPDTNYLFMGDYVDRGYYSVETVTLLVALKVRYPERITILRGNHESRQITQVYGFYDECLRKYGNANVWKYFTDLFDYLPLTALVDGQIFCLHGGLSPSIDTLDHIRALDRLQEVPHEGPMCDLLWSDPDDRGGWGISPRGAGYTFGQDISETFNHANGLTLVSRAHQLVMEGYNWCHDRNVVTIFSAPNYCYRCGNQAAIMELDDTLKYSFLQFDPAPRRGEPHVTRRTPDYFL | |
| 0.1 mL | |
| Wnt Signaling Pathway | |
| 5516 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction