missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PPP1R9B Monoclonal Antibody (K94019_12A3), Invitrogen™

Mouse Monoclonal Antibody

£380.00 - £570.00

Specifications

Antigen PPP1R9B
Clone K94019_12A3
Concentration 1 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30201781
View Documents
Invitrogen™
MA555632
50 μL
£380.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30207538
View Documents
Invitrogen™
MA556012
200 μL
£570.00
200µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sequence of this protein is as follows: FSTAPIQVFS TYSNEDYDRR NEDVDPMAAS AEYELEKRVE RLELFPVELE KDSEGLGISI IGMGAGADMG LEKLGIFVKT VTEGGAAHRD GRIQVNDLLV EVDGTSLVGV TQSFAASVLR NTKGRVRFMI GRERPGEQSE VAQLIQQTLE QERWQREMME QRYAQYGEDD EETGEYATDE DEELSPTFPG GEMAIEVFEL AENEDALSPV DMEPEKLVHK FKELQIKHAV TEAEIQQLKR KLQSLEQEKG RWRVEKAQLE QSVEENKERM EKLEGYWGEA QSLCQAVDEH LRETQAQYQA LERKYSKAKR LIKDYQQKEI EFLKKETAQR RVLEESELAR KEEMDKLLDK ISELEGNLQT

PPP1R9B Monoclonal antibody specifically detects PPP1R9B in Human, Rat samples. It is validated for Western Blot
Specifications

Specifications

PPP1R9B
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Neb2; neurabin 2; neurabin II; neurabin-2; Neurabin-II; neural tissue-specific F-actin-binding protein II; nuerabin 2; p130; PP1bp134; PPP1R6; PPP1R9; PPP1R9B; Protein phosphatase 1 regulatory subunit 9B; protein phosphatase 1, regulatory (inhibitor) subunit 9B; protein phosphatase 1, regulatory subunit 9B; protein phosphatase 1, regulatory subunit 9B, spinophilin; SPINO; Spinophilin; SPL; Spn
Recombinant human PPP1R9B (immunogen range is 451-810).
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Ppp1r9b
K94019_12A3
Western Blot
Unconjugated
Mouse
Human, Rat
O35274, Q96SB3
84686, 84687
IgG1
Protein A
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.