missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPP1R9A Polyclonal Antibody

Description
Positive Control: Rat brain tissue, SH-SY5Y, human liver carcinoma tissue, mouse brain tissue. Subcellular Location: Cell junction, Cytoplasm, Cytoskeleton, Synapse, Synaptosome.
Neurabin I is a brain-specific protein which contains an F-actin binding domain, a PDZ domain, a transmembrane-protein-interacting-domain, and a coiled-coil region. As its structure suggests, neurabin I is a promiscuous protein binding to F-actin, protein phosphatase 1, TGN38, and p70 S6 kinase. Found exclusively in brain, this protein is highly concentrated in the synapse and has also been shown to be enriched in the lamellipodia of the growth cone during neuronal development. Neurabin I appears to be a bridging protein by targeting other proteins to the synapse or by linking membrane proteins to the actin cytoskeleton.
Specifications
Specifications
| Antigen | PPP1R9A |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | PPP1R9A |
| Gene Accession No. | O35867, Q9ULJ8 |
| Gene Alias | 2810430P21Rik; 4930518N04Rik; 5330407E15; A230094E16Rik; actin-binding protein neurabin; BB181831; KIAA1222; mKIAA1222; Neb1; neurabin 1; Neurabin I; neurabin-1; Neurabin-I; neural tissue-specific F-actin binding protein; neural tissue-specific F-actin-binding protein I; NRB; NRB1; NRBI; p180; PP1bp175; Ppp1r9a; Protein phosphatase 1 regulatory subunit 9A; protein phosphatase 1, regulatory (inhibitor) subunit 9A; protein phosphatase 1, regulatory subunit 9A |
| Gene Symbols | PPP1R9A |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?