missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPP1R7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA544565
This item is not returnable.
View return policy
Description
Peptide sequence: QNLIKCIENL EELQSLRELD LYDNQIKKIE NLEALTELEI LDISFNLLRN Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
In eukaryotes, the phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions, including division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the protein phosphatases. In general, the protein phosphatase 1 (PP1) holoenzyme is a trimeric complex composed of a regulatory subunit, a variable subunit, and a catalytic subunit. Sds22, also known as PPP1R7 (protein phosphatase 1, regulatory (inhibitor) subunit 7), is a 360 amino acid protein that localizes to the nucleus and contains ten LRR (leucine rich) repeats. Expressed in a variety of tissues, Sds22 functions as a regulatory subunit of the PP1 complex, suggesting a role in protein regulation throughout the cell. Multiple isoforms of Sds22 exist due to alternative splicing events.
Specifications
| PPP1R7 | |
| Polyclonal | |
| Unconjugated | |
| PPP1R7 | |
| 2310014J01Rik; hypothetical protein LOC560323; im:7136431; Ppp1r7; Protein phosphatase 1 regulatory subunit 22; protein phosphatase 1 regulatory subunit 7; protein phosphatase 1, regulatory (inhibitor) subunit 7; protein phosphatase 1, regulatory subunit 7; protein phosphatase-1 regulatory subunit 7; Sds22; testis secretory sperm-binding protein Li 210a; zgc:123325 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5510 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q15435 | |
| PPP1R7 | |
| Synthetic peptide directed towards the middle region of human PPP1R7. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction