missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP1R7 Antibody (OTI4F9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47924
This item is not returnable.
View return policy
Description
PPP1R7 Monoclonal specifically detects PPP1R7 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| PPP1R7 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:50-100, Immunohistochemistry-Paraffin 1:50 | |
| PPP1R7 | |
| Full length human recombinant protein of human PPP1R7 (NP_002703) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| 5510 | |
| Human, Mouse, Rat, Canine | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI4F9 | |
| Unconjugated | |
| Protein phosphatase 1 regulatory subunit 22, protein phosphatase 1 regulatory subunit 7, protein phosphatase 1, regulatory (inhibitor) subunit 7, protein phosphatase-1 regulatory subunit 7 alpha2, protein phosphatase-1 regulatory subunit 7 beta1, protein phosphatase-1 regulatory subunit 7 beta2, regulatory subunit 7, sds22 | |
| Mouse | |
| 41.4 kDa | |
| 0.1 mL | |
| Primary | |
| This antibody is specific for Homo sapiens protein phosphatase 1, regulatory (inhibitor) subunit 7 (PPP1R7). | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction