missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ PPP1R3C Antibody, Novus Biologicals™

Product Code. 18345919 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18345919 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18345919 Supplier Novus Biologicals™ Supplier No. H00005507B02P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

PPP1R3C Polyclonal antibody specifically detects PPP1R3C in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R3C
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_005389.1
Gene Alias Phosphatase 1, regulatory inhibitor subunit 5, PPP1R5PP1 subunit R5, protein phosphatase 1 regulatory subunit 3C, Protein phosphatase 1 regulatory subunit 5, protein phosphatase 1, regulatory (inhibitor) subunit 3C, Protein targeting to glycogen, PTG
Host Species Mouse
Immunogen PPP1R3C (NP_005389.1, 1 a.a. - 317 a.a.) full-length human protein. MSCTRMIQVLDPRPLTSSVMPVDVAMRLCLAHSPPVKSFLGPYDEFQRRHFVNKLKPLKSCLNIKHKAKSQNDWKCSHNQAKKRVVFADSKGLSLTAIHVFSDLPEEPAWDLQFDLLDLNDISSALKHHEEKNLILDFPQPSTDYLSFRSHFQKNFVCLENCSLQERTVTGTVKVKNVSFEKKVQIRITFDSWKNYTDVDCVYMKNVYGGTDSDTFSFAIDLPPVIPTEQKIEFCISYHANGQVFWDNNDGQNYRIVHVQWKPDGVQTQMAPQDCAFHQTSPKTELESTIFGSPRLASGLFPEWQSWGRMENLASYR
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5507
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.