missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP1R3B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27285-1-AP-150UL
This item is not returnable.
View return policy
Description
Acts as a glycogen-targeting subunit for phosphatase PP1. Facilitates interaction of the PP1 with enzymes of the glycogen metabolism and regulates its activity. Suppresses the rate at which PP1 dephosphorylates (inactivates) glycogen phosphorylase and enhances the rate at which it activates glycogen synthase and therefore limits glycogen breakdown. Its activity is inhibited by PYGL, resulting in inhibition of the glycogen synthase and glycogen phosphorylase phosphatase activities of PP1. Dramatically increases basal and insulin-stimulated glycogen synthesis upon overexpression in hepatocytes.Specifications
| PPP1R3B | |
| Polyclonal | |
| Unconjugated | |
| PPP1R3B | |
| GL, PP1 subunit R4, PPP1R3B, PPP1R4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 244416, 79660 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q86XI6, Q8C767 | |
| PPP1R3B | |
| PPP1R3B Fusion Protein Ag26025 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction