missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPP1R3B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585685
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Mouse (82%), Rat (87%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Acts as a glycogen-targeting subunit for phosphatase PP1. Facilitates interaction of the PP1 with enzymes of the glycogen metabolism and regulates its activity. Suppresses the rate at which PP1 dephosphorylates (inactivates) glycogen phosphorylase and enhances the rate at which it activates glycogen synthase and therefore limits glycogen breakdown. Its activity is inhibited by PYGL, resulting in inhibition of the glycogen synthase and glycogen phosphorylase phosphatase activities of PP1. Dramatically increases basal and insulin-stimulated glycogen synthesis upon overexpression in hepatocytes.
Specifications
| PPP1R3B | |
| Polyclonal | |
| Unconjugated | |
| PPP1R3B | |
| 33 kDa glycogen-binding protein; 6430576E21; AW821953; GL; Hepatic glycogen-targeting protein phosphatase 1 regulatory subunit GL; hepatic glycogen-targeting subunit, G(L); PP1 subunit R4; PPP1R3B; PPP1R4; protein phosphatase 1 (GL-subunit); protein phosphatase 1 regulatory subunit 3B; Protein phosphatase 1 regulatory subunit 4; protein phosphatase 1 subunit GL; protein phosphatase 1, regulatory (inhibitor) subunit 3B; protein phosphatase 1, regulatory subunit 3B; PTG; R4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79660 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1.57 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q86XI6 | |
| PPP1R3B | |
| Recombinant protein encompassing a sequence within the center region of human PPP1R3B. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction